missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPD52L2/D54 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36548IR
This item is not returnable.
View return policy
Description
TPD52L2/D54 Polyclonal antibody specifically detects TPD52L2/D54 in Human samples. It is validated for Western Blot
Specifications
| TPD52L2/D54 | |
| Polyclonal | |
| Western Blot | |
| DKFZp686A1765, HCCR-binding protein 2, tumor protein D52-like 2hD54D54, tumor protein D54 | |
| The immunogen recognized by the antibody maps to a region between residue 156 to 206 of human Tumor Protein D52-like 2 using the numbering given in entry NP_003279.2. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| 7165 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction