missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPD52 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-06090
This item is not returnable.
View return policy
Description
TPD52 Polyclonal specifically detects TPD52 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| TPD52 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| D52, hD52, N8L, PC-1, PrLZ, prostate and colon associated protein, prostate leucine zipper, Protein N8, tumor protein D52 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize all three reported isoforms (NP_001020423.1, NP_001020424.1and NP_005070.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_001020423.1 | |
| TPD52 | |
| Peptide with sequence C-TEPLPEKTQES corresponding to C-Terminus according to NP_001020423.1 NP_001020424.1, NP_005070.1. | |
| 0.1 mg | |
| Cancer | |
| 7163 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction