missing translation for 'onlineSavingsMsg'
Learn More

TPD52 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35112-20ul

 View more versions of this product

Product Code. 30231330

  • £164.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TPD52 Polyclonal antibody specifically detects TPD52 in Human,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TPD52
Polyclonal
Western Blot 1:1000 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
D52, hD52, N8L, PC-1, PrLZ, prostate and colon associated protein, prostate leucine zipper, Protein N8, tumor protein D52
Recombinant fusion protein containing a sequence corresponding to amino acids 1-184 of human TPD52 (NP_005070.1).,, Sequence:, MDRGEQGLLRTDPVPEEGEDVAATISATETLSEEEQEELRRELAKVEEEIQTLSQVLAAKEKHLAEIKRKLGINSLQELKQNIAKGWQDVTATSAYKKTSETLSQAGQKASAAFSSVGSVITKKLEDVKNSPTFKSFEEKVENLKSKVGGTKPAGGDFGEVLNSAANASATTTEPLPEKTQESL
20 μL
Cancer
7163
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.