missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPCN1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£199.00 - £373.00
Specifications
| Antigen | TPCN1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TPCN1 Polyclonal antibody specifically detects TPCN1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| TPCN1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 53373 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| FLJ20612, KIAA1169, TPC1, two pore segment channel 1, two-pore channel 1, two-pore segment channel 1, voltage-dependent calcium channel protein TPC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 692-816 of human TPCN1 (NP_060371.2). FRMNYSRKNQDSEVDGGITLEKEISKEELVAVLELYREARGASSDVTRLLETLSQMERYQQHSMVFLGRRSRTKSDLSLKMYQEEIQEWYEEHAREQEQQRQLSSSAAPAAQQPPGSRQRSQTVT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title