missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TPCA-1

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Potent, selective inhibitor of IκB kinase (IKK) β (IC50 = 17.9 nM) that displays > 22-fold selectivity over IKKα and > 550-fold selectivity over other kinases and enzymes. Inhibits production of pro-inflammatory cytokines in vitro and in vivo and inhibits NF-κB nuclear localization. Reduces the severity and onset of collagen-induced arthritis; anti-inflammatory.
Specifications
Specifications
| CAS | 507475-17-4 |
| Quantity | 10 mg |
| Molecular Formula | C12H10FN3O2S |
| Formula Weight | 279.29 |
| Purity | >97% |
| Molecular Weight (g/mol) | 279.289 |
| Chemical Name or Material | TPCA-1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction