missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TP73-AS1 Monoclonal Antibody (OTI2B9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525703
This item is not returnable.
View return policy
Description
TP73-AS1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
TP73-AS1 in molecular biology is P73 antisense RNA 1T (non-protein coding) and also known as PDAM, is a long non-coding RNA. It may regulate apoptosis via regulation of p53-dependent anti-apoptotic genes. It may play a role in the development of oligodendroglial tumours.
Specifications
| TP73-AS1 | |
| Monoclonal | |
| 0.65 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 0 | |
| TP73-AS1 | |
| Full length human recombinant protein of KIAA0495. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2B9 | |
| Unconjugated | |
| TP73-AS1 | |
| KIAA0495; PDAM; TP73 antisense RNA 1; TP73-AS1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57212 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction