missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TP63 Recombinant Rabbit Monoclonal Antibody (10H7L17)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703809
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Monkey, Mouse, Rat, Bovine.
TP63 (tumor protein 63) acts as a sequence-specific DNA binding transcriptional activator or repressor. The isoforms contain a varying set of transactivation and auto-regulating transactivation inhibiting domains thus showing an isoform-specific activity. Isoform 2 activates RIPK4 transcription. TP63 may be required in conjunction with TP73/p73 for initiation of p53/TP53 dependent apoptosis in response to genotoxic insults and the presence of activated oncogenes. It is involved in Notch signaling by inducing JAG1 and JAG2. TP63 plays a role in the regulation of epithelial morphogenesis. The ratio of DeltaN-type and TA*-type isoforms may govern the maintenance of epithelial stem cell compartments and regulate the initiation of epithelial stratification from the undifferentiated embryonal ectoderm. TP63 is required for limb formation from the apical ectodermal ridge. It also activates transcription of the p21 promoter. Mutations affecting the gene can result in Acro-dermato-ungual-lacrimal-tooth syndrome, ankyloblepharon-ectodermal defects-cleft lip/palate, ectrodactyly ectodermal dysplasia, split-hand/foot malformation 4, limb-mammary syndrome, ectodermal dysplasia - Rapp-Hodgkin type, and orofacial cleft 8.
Specifications
| TP63 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9H3D4 | |
| TP63 | |
| Protein corresponding to human TP63 [aa360-aa500]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry, Western Blot | |
| 10H7L17 | |
| Unconjugated | |
| TP63 | |
| AIS; amplified in squamous cell carcinoma; B(p51A); B(p51B); Chronic ulcerative stomatitis protein; CUSP; EEC3; Keratinocyte transcription factor KET; KET; LMS; NBP; OFC8; p40; p51; p53CP; p63; p63a; P73H; p73L; RHS; SHFM4; TP53CP; TP53L; TP63; TP73L; Transformation-related protein 63; tumor protein 63; tumor protein p53-competing protein; tumor protein p63; Tumor protein p73-like | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8626 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction