missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TP53I13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569780
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Guinea Pig: 79%; Horse: 85%; Human: 100%; Mouse: 79%; Pig: 86%; Rabbit: 79%; Rat: 79%.
TP53I13 gene ontology annotations related to this gene include negative regulation of cell cycle; response to drug; response to organic cyclic compound; response to UV.
Specifications
| TP53I13 | |
| Polyclonal | |
| Unconjugated | |
| TP53I13 | |
| 2410019G02Rik; Damage-stimulated cytoplasmic protein 1; DSCP1; LOW QUALITY PROTEIN: tumor protein p53-inducible protein 13; RGD1559818; Tp53i13; transformation related protein 53 inducible protein 13; Trp53i13; tumor protein p53 inducible protein 13; tumor protein p53-inducible protein 13 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 90313 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NBR0 | |
| TP53I13 | |
| synthetic peptide directed towards the middle region of human TP53I13. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction