missing translation for 'onlineSavingsMsg'
Learn More

TP0463518, MedChemExpress

Product Code. 16400887
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400887

Brand: Medchem Express HY112144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TP0463518 is a potent hypoxia-inducible factor prolyl hydroxylases (PHDs) inhibitor with a Ki value of 5.3 nM for human PHD2. TP0463518 also inhibits human PHD1/PHD3 with IC50s of 18 and 63 nM as well as monkey PHD2 with an IC50 value of 22 nM.

TP0463518 is a potent hypoxia-inducible factor prolyl hydroxylases (PHDs) inhibitor with a Ki value of 5.3 nM for human PHD2. TP0463518 also inhibits human PHD1/PHD3 with IC50s of 18 and 63 nM as well as monkey PHD2 with an IC50 value of 22 nM.

Specifications

Chemical Name or Material TP0463518
CAS 1558021-37-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H18ClN3O6
Quantity 50 mg
Solubility Information DMSO : ≥ 125 mg/mL (289.47 mM)
SMILES ClC(C=C1)=CC=C1OC2=NC=C(CN3C(C(C(NCC(O)=O)=O)=C(O)CC3)=O)C=C2
Molecular Weight (g/mol) 431.83
Formula Weight 431.83
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.