missing translation for 'onlineSavingsMsg'
Learn More

TP0463518, MedChemExpress

Product Code. 16484578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484578 10 mg 10mg
16437297 5 mg 5mg
16405436 100 mg 100mg
16400887 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484578 Supplier Medchem Express Supplier No. HY112144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TP0463518 is a potent hypoxia-inducible factor prolyl hydroxylases (PHDs) inhibitor with a Ki value of 5.3 nM for human PHD2. TP0463518 also inhibits human PHD1/PHD3 with IC50s of 18 and 63 nM as well as monkey PHD2 with an IC50 value of 22 nM.

TP0463518 is a potent hypoxia-inducible factor prolyl hydroxylases (PHDs) inhibitor with a Ki value of 5.3 nM for human PHD2. TP0463518 also inhibits human PHD1/PHD3 with IC50s of 18 and 63 nM as well as monkey PHD2 with an IC50 value of 22 nM.

Specifications

Chemical Name or Material TP0463518
CAS 1558021-37-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H18ClN3O6
Quantity 10 mg
Solubility Information DMSO : ≥ 125 mg/mL (289.47 mM)
SMILES ClC(C=C1)=CC=C1OC2=NC=C(CN3C(C(C(NCC(O)=O)=O)=C(O)CC3)=O)C=C2
Molecular Weight (g/mol) 431.83
Formula Weight 431.83
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.