missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toxoplasma Gondii P30 Antibody (P30/3), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66486MFV610
This item is not returnable.
View return policy
Description
Toxoplasma Gondii P30 Monoclonal antibody specifically detects Toxoplasma Gondii P30 in Protozoa, T. gondii samples. It is validated for Flow Cytometry
Specifications
| Toxoplasma Gondii P30 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Protozoa, T. gondii | |
| Purified |
| Flow Cytometry | |
| P30/3 | |
| Flow Cytometry | |
| p30, SAG1, SAG1 protein | |
| Purified T. gondii P30 antigen | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction