missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toxoplasma Gondii P30 Antibody (P30/3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66486AF700
This item is not returnable.
View return policy
Description
Toxoplasma Gondii P30 Monoclonal antibody specifically detects Toxoplasma Gondii P30 in Protozoa, T. gondii samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Toxoplasma Gondii P30 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| p30, SAG1, SAG1 protein | |
| Purified T. gondii P30 antigen | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| P30/3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Protozoa, T. gondii | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction