missing translation for 'onlineSavingsMsg'
Learn More

Toxicarol isoflavone, MedChemExpress

Product Code. 16492559
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492559 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492559 Supplier Medchem Express Supplier No. HYN1135
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Toxicarol isoflavone is an isoflavone extracted from Millettia brandisiana.

Toxicarol isoflavone is an isoflavone extracted from Millettia brandisiana.

Specifications

Chemical Name or Material Toxicarol isoflavone
CAS 3044-60-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H22O7
Quantity 1 mg
SMILES O=C1C2=C(O)C=C3C(C=CC(C)(C)O3)=C2OC=C1C4=CC(OC)=C(OC)C=C4OC
Molecular Weight (g/mol) 410.42
Formula Weight 410.42
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.