missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ToxBLAzer™ DualScreen Kit

Product Code. 11504936
Click to view available options
Quantity:
200 μg
5mg
Unit Size:
5mg
Each
This item is not returnable. View return policy

Product Code. 11504936

Brand: Invitrogen™ K1139

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ToxBLAzer™ DualScreen Kit

ToxBLAzer™ DualScreen Kit provide the key advantage of combining the ratiometric reporter gene readout of beta-lactamase with a cytotoxicity readout in the same assay. Thus, ToxBLAzer™ DualScreen yields reporter readouts that normalize cytotoxic cellular responses, which is a necessity in screening applications which down regulate transcription. The ToxBLAzer™ substrate contains both the LiveBLAzer™-FRET B/G substrate and a proprietary cytotoxicity indicator. Both compounds readily enter the cell, where endogenous esterases rapidly convert them into fluorescent compounds Esterase activity and an intact cell membrane are required for detection of the indicator. The cytotoxicity indicator's fluorescence is red-shifted to ensure compatibility with the blue and green emissions of the LiveBLAzer™-FRET B/G substrate, allowing detection of both reporter gene activity and cytotoxicity in the same assay sample.

Advantages of ToxBLAzer™ DualScreen Kits:

  • Multiplexed readout results in rapid identification of false positives due to toxicity or lack of viable cells, thereby improving hit-picking efficiency
  • Ratiometric blue:green reporter readout reduces false hits by minimizing experimental noise
  • Single reagent addition eliminates need for follow up cytotoxicity assays, thereby increasing throughput while reducing sample and test compound consumption
  • TRUSTED_SUSTAINABILITY

    Specifications

    Formulation Liquid
    For Use With (Application) Protein biology, cell analysis, cellular imaging, cell-based reporter assays, cell-based and beta-lactamase assays, cell viability, proliferation and function, cell viability and cytotoxicity, drug discovery and development, target and lead identification and validation, kinase biology, kinase pathway analysis, G-protein coupled receptor biology, cell-based GPCR reporter assays, nuclear receptor biology, cell-based nuclear receptor reporter assays, pathway biology, cellular pathway analysis assays, other cellular pathway assay products, enzyme reporter assays, protein assays and analysis, beta-lactamase activity assays
    Includes ToxBLAzer™ substrate, DMSO (for solution A), solution B, and solution C. ToxBLAzer™ substrate, DMSO (for solution A), solution B, and solution C should
    pH Range 6 to 8
    Product Type ToxBLAzer™ DualScreen Kit
    Quantity 5mg

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.