missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555653
This item is not returnable.
View return policy
Description
Immunogen sequence: PMATVDPASP APASIEPPAL SPSIVVNSTL SSYVANQASS GAGGQPNITK LIITKQMLPS SITMSQGGMV TVIPATVVTS RGLQLGQTST ATIQPSQQAQ IVTRSVLQAA AAAAAAASMQ LPPPRLQPPP LQQMPQPPTQ QQVTI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 94%.
KIAA0737 contains 1 HMG box DNA-binding domain and the function remains unknows.
Specifications
| TOX4 | |
| Polyclonal | |
| Unconjugated | |
| TOX4 | |
| 5730589K01Rik; A630040M18; AA410149; C14orf92; Epidermal Langerhans cell protein LCP1; KIAA0737; LCP1; MIG7; migration-inducing protein 7; RGD708449; TOX high mobility group box family member 4; TOX4; Unknown (protein for MGC:139878) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9878 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94842 | |
| TOX4 | |
| Recombinant protein corresponding to Human TOX4. Recombinant protein control fragment (Product #RP-88895). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction