missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOX3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15767011 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15767011 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15767011 Supplier Invitrogen™ Supplier No. PA559151

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SQGSEFTPQF PPQSLDLPSI TISRNLVEQD GVLHSSGLHM DQSHTQVSQY RQDPSLIMRS IVHMTDAARS GVMP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

TOX3 contains an HMG-box, indicating that it may be involved in bending and unwinding of DNA and alteration of chromatin structure. The C-terminus of the encoded protein is glutamine-rich due to CAG repeats in the coding sequence. A minor allele of this gene has been implicated in an elevated risk of breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOX3
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOX3
Gene Accession No. O15405
Gene Alias 500-9; BC052044; C230068E13; CAG trinucleotide repeat-containing gene F9 protein; CAGF9; Tnrc9; TOX high mobility group box family member 3; TOX3; trinucleotide repeat containing 9; trinucleotide repeat-containing gene 9 protein
Gene Symbols TOX3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOX3. Recombinant protein control fragment (Product #RP-98363).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27324
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.