missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOX3 Antibody (TOX3/1124), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-45166-0.1MG
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunofluorescence
TOX3 Monoclonal specifically detects TOX3 in Human samples. It is validated for Western Blot, ELISA.
Specifica
| TOX3 | |
| Monoclonal | |
| 0.2mg/mL | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| O15405 | |
| TOX3 | |
| Recombinant fragment (139 Amino acid residues around aa 200-400) of human TOX3 protein | |
| Protein A or G purified | |
| RUO | |
| 27324 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, SDS-Page, Immunofluorescence | |
| TOX3/1124 | |
| Unconjugated | |
| 1.0mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CAG trinucleotide repeat-containing gene F9 protein, CAGF9trinucleotide repeat containing 9, TNRC9, TOX high mobility group box family member 3, Trinucleotide repeat-containing gene 9 protein | |
| Mouse | |
| 63 kDa | |
| 0.1 mg | |
| Primary | |
| It recognizes a 63kDa protein, which is identified as TOX3. It contains a high mobility group (HMG)-box, which regulates Ca2+-dependent transcription in neurons through interaction with the cAMP-response-element-binding protein (CREB). TOX3 appears to be associated with breast cancer susceptibility and is expressed downstream of a cytoprotective cascade together with CITED1, a transcriptional regulator that does not bind directly to DNA. TOX3 is predominantly expressed in the brain and forms homodimers. TOX3 overexpression protects neuronal cells from cell death caused by endoplasmic reticulum stress or BAX overexpression through the induction of anti-apoptotic transcripts and repression of pro-apoptotic transcripts. | |
| Store at 4C. | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto