missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOX3 Antibody (TOX3/1123), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47748R
This item is not returnable.
View return policy
Description
TOX3 Monoclonal antibody specifically detects TOX3 in Human, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications
| TOX3 | |
| Monoclonal | |
| DyLight 550 | |
| CAG trinucleotide repeat-containing gene F9 protein, CAGF9trinucleotide repeat containing 9, TNRC9, TOX high mobility group box family member 3, Trinucleotide repeat-containing gene 9 protein | |
| Recombinant fragment of human TOX3 protein (around aa 251-389) (Exact sequence is proprietary) (Uniprot: O15405) | |
| 0.1 mL | |
| Primary | |
| Human, Chicken | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| TOX3/1123 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 27324 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction