missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOX2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21162-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene, which is located on chromosome 20, is predicted to encode for a signal protein with unknown function.Specifications
| TOX2 | |
| Polyclonal | |
| Unconjugated | |
| TOX2 | |
| C20orf100, GCX 1, GCX1, TOX2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 269389, 84969 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96NM4 | |
| TOX2 | |
| TOX2 Fusion Protein Ag14772 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction