Learn More
Invitrogen™ TOX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144032
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat thymus tissue, rat liver tissue, mouse thymus tissue, mouse liver tissue. ICC/IF: A549 cell. Flow: MCF-7 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
TOX2 (TOX High Mobility Group Box Family Member 2) is a Protein Coding gene. Diseases associated with TOX2 include Conduct Disorder. Gene Ontology (GO) annotations related to this gene include chromatin binding and RNA polymerase II transcription factor binding. An important paralog of this gene is TOX.
Specifications
| TOX2 | |
| Polyclonal | |
| Unconjugated | |
| TOX2 | |
| AI851523; AV026525; BI987407; C20orf100; dJ1108D11.2; dJ495O3.1; GCX1; Gcx-1; granulosa cell HMG box 1; granulosa cell HMG box protein 1; granulosa cell HMG-box protein 1; granulosa cell HMG-box protein-1; LOW QUALITY PROTEIN: TOX high mobility group box family member 2; RxHMG1; TOX high mobility group box family member 2; TOX2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 269389, 311615, 84969 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q76IQ7, Q96NM4 | |
| TOX2 | |
| E.coli-derived human TOX2 recombinant protein (Position: Q41-A425). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.