Learn More
TOX2 Mouse, anti-Human, Clone: 2G12A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68590-1-IG-150UL
Description
The TOX2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TOX2. This antibody recognizes human, mouse, rat, rabbit antigen. The TOX2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TOX2 | |
| 2G12A7 | |
| C20orf100, dJ1108D11.2, dJ495O3.1, GCX 1, GCX1, TOX2 | |
| 150 μL | |
| 84969 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| 150 kDa | |
| IgG1 |