missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534423
This item is not returnable.
View return policy
Description
A suggested positive control is human colon tissue lysate. PA5-34423 can be used with blocking peptide PEP-1465.
TOX (thymocyte selection-associated high mobility group (HMG) box protein) is a member of the HMG box family of DNA-binding proteins and likely plays a role in the regulation of T-cell development. It is a 526 amino acid nuclear protein and the expression of TOX is upregulated by pre-T cell receptor (pre-TCR) and TCR activation in immature thymocytes. TOX-mediated positive selection is associated with up-regulation of RUNX3 and is calcineurin dependent. TOX-dependent transition to the CD4+CD8 stage is required for development of class II major histocompatibility complex-specific T cells.
Specifications
| TOX | |
| Polyclonal | |
| Unconjugated | |
| TOX | |
| 1700007F02Rik; KIAA0808; thymocyte selection associated high mobility group box; thymocyte selection-associated high mobility group box; thymocyte selection-associated high mobility group box protein TOX; thymocyte selection-associated HMG box; thymus high mobility group box protein TOX; TOX; TOX1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 252838, 9760 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O94900, Q66JW3 | |
| TOX | |
| A 17 amino acid synthetic peptide near the amino terminus of human TOX. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction