Learn More
Invitrogen™ TOX Monoclonal Antibody (TXRX10), eFluor™ 660, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£137.00 - £334.00
Specifications
| Antigen | TOX |
|---|---|
| Clone | TXRX10 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15500007
|
Invitrogen™
50-6502-80 |
25 μg |
£137.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15510007
![]() |
Invitrogen™
50-6502-82 |
100 μg |
£334.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This TXRX10 monoclonal antibody reacts with human and mouse TOX, which is also known as Thymocyte selection-associated HMG box protein. This DNA-binding nuclear factor is expressed in thymocytes, T lymphocytes, NK cells, and lymphoid tissue-inducer (LTi) cells. TOX expression in thymocytes is upregulated during beta-selection and positive selection. Studies using Tox knockout mice have demonstrated a critical role for TOX in CD4+ T cell development as a regulator of the CD4loCD8lo to CD4+CD8lo transition. Moreover, TOX has been shown to be involved in the development of NK and LTi cells. While its regulation remains unclear, TOX expression has been shown to be induced by calcineurin-mediated TCR signaling in thymocytes during positive selection. Applications Reported: This TXRX10 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This TXRX10 antibody has been tested by intracellular staining and flow cytometric analysis of mouse thymocytes using the Foxp3/Transcriptin Factor Buffer Set (cat 00-5523) and protocol. Refer to Best Protocols- (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
TOX (thymocyte selection-associated high mobility group (HMG) box protein) is a member of the HMG box family of DNA-binding proteins and likely plays a role in the regulation of T-cell development. It is a 526 amino acid nuclear protein and the expression of TOX is upregulated by pre-T cell receptor (pre-TCR) and TCR activation in immature thymocytes. TOX-mediated positive selection is associated with up-regulation of RUNX3 and is calcineurin dependent. TOX-dependent transition to the CD4+CD8 stage is required for development of class II major histocompatibility complex-specific T cells.Specifications
| TOX | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 1700007F02Rik; KIAA0808; thymocyte selection associated high mobility group box; thymocyte selection-associated high mobility group box; thymocyte selection-associated high mobility group box protein TOX; thymocyte selection-associated HMG box; thymus high mobility group box protein TOX; TOX; TOX1 | |
| TOX | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| TOX |
| TXRX10 | |
| Flow Cytometry | |
| eFluor 660 | |
| Rat | |
| Human, Mouse | |
| O94900, Q66JW3 | |
| 252838, 9760 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.