Learn More
Invitrogen™ TOX Monoclonal Antibody (TXRX10), eBioscience™

Rat Monoclonal Antibody
£129.00 - £3445.00
Specifications
| Antigen | TOX |
|---|---|
| Clone | TXRX10 |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C |
| Applications | Immunoprecipitation, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17198525
|
Invitrogen™
14-6502-37 |
2 mg |
£1392.00
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15237717
|
Invitrogen™
14-6502-80 |
25 μg |
£129.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15247717
![]() |
Invitrogen™
14-6502-82 |
100 μg |
£263.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17108535
|
Invitrogen™
14-6502-95 |
10 mg |
£3445.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This TXRX10 monoclonal antibody reacts with human and mouse TOX, which is also known as Thymocyte selection-associated HMG box protein. This DNA-binding nuclear factor is expressed in thymocytes, T lymphocytes, NK cells, and lymphoid tissue-inducer (LTi) cells. TOX expression in thymocytes is upregulated during beta-selection and positive selection. Studies using Tox knockout mice have demonstrated a critical role for TOX in CD4^+ T cell development as a regulator of the CD4^loCD8^lo to CD4^+CD8^lo transition. Moreover, TOX has been shown to be involved in the development of NK and LTi cells. While its regulation remains unclear, TOX expression has been shown to be induced by calcineurin-mediated TCR signaling in thymocytes during positive selection. Applications Reported: This TXRX10 antibody has been reported for use in immunoprecipitation and immunoblotting (WB). Applications Tested: This TXRX10 antibody has been tested by immunoblotting of reduced cell lysate prepared from mouse thymocytes at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
TOX (thymocyte selection-associated high mobility group (HMG) box protein) is a member of the HMG box family of DNA-binding proteins and likely plays a role in the regulation of T-cell development. It is a 526 amino acid nuclear protein and the expression of TOX is upregulated by pre-T cell receptor (pre-TCR) and TCR activation in immature thymocytes. TOX-mediated positive selection is associated with up-regulation of RUNX3 and is calcineurin dependent. TOX-dependent transition to the CD4+CD8 stage is required for development of class II major histocompatibility complex-specific T cells.Specifications
| TOX | |
| 0.5 mg/mL | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| O94900, Q66JW3 | |
| 252838, 9760 | |
| Primary | |
| Affinity chromatography |
| TXRX10 | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human, Mouse | |
| TOX | |
| 1700007F02Rik; KIAA0808; thymocyte selection associated high mobility group box; thymocyte selection-associated high mobility group box; thymocyte selection-associated high mobility group box protein TOX; thymocyte selection-associated HMG box; thymus high mobility group box protein TOX; TOX; TOX1 | |
| TOX | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.