Learn More
Brady™ ToughBond™ B-422 Polyester BMP71 Printer Labels with Permanent Adhesive
For use in commercial/industrial applications. Brady B-422 polyester label are for use with BMP71 printer.
£44.95 - £123.00
Specifications
Certifications/Compliance UL Recognized; CSA Accepted
Description Chemical, Corrosion, Humidity, UV and Weather-Resistant
Adhesive Type Acrylic, Permanent
Color White
Finish Gloss
View More Specs
Products 9
Product Code Brand Size (Metric) Size (English) Quantity Price Quantity & Availability  
Product Code Brand Size (Metric) Size (English) Quantity Price Quantity & Availability  
11543924
View Documents Promotion Details
Brady™
114707
W x H: 25.4 mm x 15.2m 50 ft. x 1 in. 1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11593964
View Documents Promotion Details
Brady™
114753
W x H: 38.1 mm x 15.2m 50 ft. x 1.5 in. 1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11548614
View Documents Promotion Details
Brady™
115138
W x H: 9.5 mm x 15.2m 50 ft. x 0.375 in. 1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11509453
View Documents Promotion Details
Brady™
104281
W x H: 12.06 mm x 15.2m 50 ft. x 0.475 in. 1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11529463
View Documents Promotion Details
Brady™
104293
W x H: 12.7 mm x 15.2m 50 ft. x 0.5 in. 1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11558614
View Documents Promotion Details
Brady™
115139
W x H: 15.8 mm x 15.2m 50 ft. x 0.625 in. 1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11727274
View Documents Promotion Details
Brady™
114803
38.1 x 19 mm (W x H) 1.5 x 0.75 in. (W x H) 250/Roll This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11767104
View Documents Promotion Details
Brady™
114822
76.2 x 48.2 mm (W x H) 3 x 1.9 in. (W x H) 100/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11751715
View Documents Promotion Details
Brady™
114833
38.1 x 19 mm (W x H) 1.5 x 0.75 in. (W x H) 500/Roll This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Specifications
UL Recognized; CSA Accepted
Acrylic, Permanent
Gloss
Asset Tracking Label
BMP™71 Printer
-40°C to +100°C
0.102 mm
Chemical, Corrosion, Humidity, UV and Weather-Resistant
White
Polyester (B-422)
Component ID, Equipment ID, Barcoding, Panel ID, Voice/Data ID, Asset tracking, Rating plates, General purpose ID
1
0.004 in.