missing translation for 'onlineSavingsMsg'
Learn More

MakerBot™ Tough PLA Filament Bundle

Product Code. 16371536
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Replicator 3D Printer, Replicator+ 3D Printer
Replicator Z18 3D Printer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment)
16371536 Replicator 3D Printer, Replicator+ 3D Printer
16381536 Replicator Z18 3D Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16371536 Supplier MakerBot™ Supplier No. MKS000082
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MakerBot™ Tough PLA Filament Bundle includes 3 spools of Tough PLA Filament and the Tough PLA Smart Extruder+.

  • Composed of the purest grade resin on the market
  • Comprehensively quality tested
  • Reduces downtime
  • Creates high quality prints

Specifications

Color Variety
For Use With (Equipment) Replicator 3D Printer, Replicator+ 3D Printer
Product Type PLA Bundle
Material PLA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.