Learn More
Bellingham and Stanley™ Touch Screen Refractometer
GREENER_CHOICE
Product Code. 16614912
Change view
Click to view available options
Description:
Resolution: 0.01 Brix
Resolution: 0.01/0.001 Brix
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
16614912 Resolution: 0.01 Brix Each
15807205 Resolution: 0.01/0.001 Brix Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16614912 Supplier Bellingham and Stanley™ Supplier No. 1930
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Peltier temperature controlled refractometer with resolution touch display
Touch Screen Refractometer with Peltier temperature control features the latest integral 7 in. full color touchscreen, high definition display, and USB/LAN connectivity. The refractometer is supplied with two RFID tags, instruction manual, and certificate of conformity. Perfect for measurement and control in demanding food, beverage, chemical and industrial applications.
  • Scales
    • RI 1.32 - 1.58
    • Brix 0 - 100
  • Resolution:
    • RI 0.00001 (RFM330-T)/ 0.000001 (RFM340-T)
    • Brix 0.01
TRUSTED_SUSTAINABILITY
Specifications
Type Digital Refractometer
Scale Range 0 to 100 Brix
Description Resolution: 0.01 Brix
Model RFM330-T
Accuracy ± 0.1 Brix
Includes Refractometer, RFID Tags (2), Instruction Manual, and Certificate of Conformity