missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ 92-600 Disposable Chemical Resistant Nitrile Gloves

Product Code. 16615135 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 100
Pack of 90
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16615135 X-Small Pack of 100
11706714 Small Pack of 100
11716584 Medium Pack of 100
11726584 Large Pack of 100
11786584 X-Large Pack of 100
32078683 2X-Large Pack of 90
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16615135 Supplier Ansell Supplier No. 92600060
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.

  • Made from 100% nitrile
  • Resistant to nuisance punctures, nicks, cuts and abrasions
  • Comfortable thin mil construction
  • Rolled cuffs for easy donning and doffing
  • Chemical resistant
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Color Green
Finger Thickness (English) 5.5 mil
Finger Thickness (Metric) 0.14 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.92 mil
Palm Thickness (Metric) 0.125 mm
Product Type Chemical-resistant Gloves
Size X-Small
Thickness (English) 4.92 mil
Thickness (Metric) 0.125 mm
Glove Cuff Style Rolled
Quantity 1000/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.