missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Tote pan rack with six tilted shelves, 24 TP3 tote pan capacity
Description
A range of heavy duty workshop racking designed for use in conjunction with Tote Pans. Strong welded steel construction in a blue painted finish. Choice of four, five, six or eight horizontal or tilted shelf levels, all units being 1350mm wide. Four tote pans per shelf level (not supplied).
- Shelf capacity UDL: 150kg
- Type: TP3
- Dimensions (H x D): 1180mm x 610mm
- Number of levels: 6
- Bin capacity: 24L
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction