missing translation for 'onlineSavingsMsg'
Learn More

Manutan Tote pan rack with eight tilted shelves, 32 TP1 tote pan capacity

Product Code. 12885668
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12885668 Supplier Manutan Supplier No. A338985

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
A range of heavy duty workshop racking designed for use in conjunction with Tote Pans. Strong welded steel construction in a blue painted finish. Choice of four, five, six or eight horizontal or tilted shelf levels, all units being 1350mm wide. Four tote pans per shelf level (not supplied).
  • Shelf capacity UDL: 150kg
  • Type: TP1
  • Dimensions (H x D): 1580mm x 305mm
  • Number of levels: 8
  • Bin capacity: 32L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.