missing translation for 'onlineSavingsMsg'
Learn More

Total Permethrin 100 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30315145
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315145 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315145 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010373AC
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Permethrin
CAS 52645-53-1
Analyte or Component Names Permethrin
Shipping Condition Room Temperature
Molecular Formula C21 H20 Cl2 O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dichloroethenyl)-2,2-dimethyl-, (3-phenoxyphenyl)methyl ester, (3-Phenoxyphenyl)methyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate, 3-Phenoxybenzyl 2,2-dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarboxylate, 3-Phenoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, Acticin, Adion, Ambush, Aninsen Per-30, Anomethrin N, Antiborer 3768, Aqua-Kontrol, Astro, Bansect, Bematin 987, BioKill, Butox 50, Chinetrin, Cooper, Coopex, Corsair, Damminix, Dancide PS 150, Dichlorophenothrin, Diffusil H, Dragnet, Dragnet FT, Dragon, Ecsumin, Ectiban, Efmethrin, Eliminator Ant, Flea & Tick Killer, Elimite, Eulan SPA, Everwood PC 30W, Exmin, FMC 33297, FMC 41655, ICI-PP 557, Imperator, InfectoPedicul, Insectal Plus, Insorbcid MP, Ipitox, JF 7065, Kaleait, Kavil, Kestrel, Kestrel (pesticide), Kudos, Last Call, Lyclear, MP 79, MP 79 (ester), Mitin BC, NIA 33297, NRDC 143, Nedax, Nix, Nix (insecticide), PP 557, Perigen, Perigen 500, Perigen Defense, Perigen W, Perm-UP, Permanone, Permanone 40, Permanone 80, Permanone 90, Permasect, Permectin II, Permethrin, Permethrin I, Permethrin II, Permethrine, Permitrine, Pertrin E, Piredam, Pounce, Preventol HS 75, Pulvex, Pynosect, Repel Permanone, Riapan A, Ridect Pour-On, S 3151, SBP 1513, SBP 15131TEC, Sharkesuper, Skintex MR-III, Smite EC 10, Spartan, Spartan (pesticide), Stomoxin, Stomozan, Tanalith T, Timbertreat ICP 5, WL 43479, m-Methoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, m-Phenoxybenzyl 3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate, GW418275X
InChI Formula InChI=1S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19H,13H2,1-2H3
SMILES CC1(C)C(C=C(Cl)Cl)C1C(=O)OCc2cccc(Oc3ccccc3)c2
IUPAC Name (3-phenoxyphenyl)methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Solvent or Matrix Acetone
Molecular Weight (g/mol) 391.29
Quantity 1 mL
Formula Weight 390.0789 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.