missing translation for 'onlineSavingsMsg'
Learn More

Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 042562.AP

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11366109

  • £190.00 / 500mL
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Total Organic Carbon (TOC), is used as a standard solution during the measurement of chemical analysis such as detecting contaminants in drinking water and cooling water. It is also used for analysis of water to be used for semiconductor manufacturing and pharmaceutical use.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Total Organic Carbon
500 mL
KHC8H4O4 in H2O
Characteristic
Liquid
1000μg/mL
Colorless
MFCD00133992
Miscible with water
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only