missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Total Nitric Oxide and Nitrate/Nitrite Parameter Assay Kit
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
96 tests
Description
The Total Nitric Oxide and Nitrate/Nitrite Parameter Assay Kit quantifies multi-species Nitric Oxide in cell culture supernates (10 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul), citrate plasma (25 ul), urine (10 ul).
Specifications
Specifications
| Assay Range | 3.12 - 200 umol/L |
| Assay Sensitivity | 0.78 umol/L |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine |
| Specificity | Total Nitric Oxide, Nitrite and Nitrate Levels in Various Samples |
| For Use With (Application) | ELISA |
| Quantity | 1 x 96-well Plate |
| Sample Volume | Cell Culture Supernates (10uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL),Citrate Plasma (25uL),Urine (10uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction