missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Total MMP-2 Quantikine ELISA Kit

Product Code. 30002143
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Unit Size:
1 set
Pack of 96
This item is not returnable. View return policy

Product Code. 30002143

Brand: R&D Systems™ SMMP200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For human, mouse, rat, porcine, and canine samples.

The Total MMP-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat, porcine, canine MMP-2 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.50 to 32 ng/mL
Assay Sensitivity 0.082 ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum, Serum
Specificity Recombinant MMP-2 and natural Human, Mouse, Rat, Porcine, and Canine active, pro-, and TIMP complexed MMP-2.
Sufficient For 6 x 96-well Plates
Target Species Canine, Human, Mouse, Porcine, Rat
For Use With (Application) ELISA
Gene ID (Entrez) 4313
Gene Symbol MMP2
Quantity 1 Pk. (6 Plates)
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72 kDa gelatinase, 72 kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72 kD gelatinase, 72 kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP2, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1 matrix metalloproteinase 2 (gelatinase A, 72 kDa gelatinase, 72 kDa type IVcollagenase)
Target MMP-2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.