missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Total Cholesterol Reagents

Product Code. 10178058 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 125 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10178058 2 x 125 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10178058 Supplier Thermo Scientific™ Supplier No. TR13421

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Determine quantity of cholesterol in serum in vitro with Thermo Scientific™ Total Cholesterol Reagents.

Measurement of serum cholesterol levels can serve as an indicator of liver, biliary or thyroid function, intestinal absorption, adrenal disease and predisposition toward coronary artery disease. Increased levels are associated with nephrosis, diabetes mellitus and obstructive jaundice. When used in conjunction with triglyceride, HDL and LDL determinations, total cholesterol assays provide valuable information for the prediction of coronary artery disease. Decreased levels may be observed in cases of hyperthyroidism, certain anemias, malabsorption and wasting syndromes.
  • Up to 12 months reconstituted stability and various kit sizes to minimize reagent wastage
  • Suitable for manual use or on automated clinical chemistry analyzers
  • Applications for a wide range of automated analyzers available on request
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Description Infinity Cholesterol 2 x 125mL
CE Marker Yes
Detectable Analytes Cholesterol
Sample Type Plasma, Serum, Serum or Plasma
Type Reagent
Linearity Up to 20mmol/L (774mg/dL)
Methods Compatibility Endpoint
Quantity 2 x 125 mL
Stability Until expiry at 2-8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.