missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Tosyl-d7)urethane, TRC
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
Specifications
Specifications
| Chemical Name or Material | (Tosyl-d7)urethane |
| Molecular Formula | C10 D7 H6 N O4 S |
| Quantity | 50 mg |
| Synonym | Ethyl (p-Toluenesulfonyl-d7)carbamate, Ethyl (p-Tolylsulfonyl-d7)carbamate, Ethyl N-(4-Methylbenzenesulfonyl-d7)carbamate, Ethyl N-(Tosyl-d7)carbamate, Ethyl p-(Tosyl-d7)carbamate, N-(Tosyl-d7)urethane, [(4-Methylphenyl-d7)sulfonyl]carbamic Acid Ethyl Ester, (p-(Tosyl-d7)sulfonyl)carbamic Acid Ethyl Ester, N-[(4-Methylphenyl-d7)sulfonyl]carbamic Acid Ethyl Ester |
| InChI Formula | InChI=1S/C10H13NO4S/c1-3-15-10(12)11-16(13,14)9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3,(H,11,12)/i2D3,4D,5D,6D,7D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1C([2H])([2H])[2H])S(=O)(=O)NC(=O)OCC |
| IUPAC Name | ethyl N-[2,3,5,6-tetradeuterio-4-(trideuteriomethyl)phenyl]sulfonylcarbamate |
| Molecular Weight (g/mol) | 250.323 |
| Formula Weight | 250.1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction