missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Torsin A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95160-0.1ml
This item is not returnable.
View return policy
Description
Torsin A Polyclonal antibody specifically detects Torsin A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Torsin A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DQ2Torsin family 1 member A, Dystonia 1 protein, dystonia 1, torsion (autosomal dominant; torsin A), DYT1torsin-1A, torsin family 1, member A (torsin A) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human TorsinA/TOR1A (NP_000104.1). PRLYCLFAECCGQKRSLSREALQKDLDDNLFGQHLAKKIILNAVFGFINNPKPKKPLTLSLHGWTGTGKNFVSKIIAENIYEGGLNSDYVHLFVATLHFPHASNITLYKDQLQLWIRGNVSACARSIFIFDEMDKMHAGLIDAIKPFLDYYDLVDGVSYQKAMFIFLSNAGAERITDVALDFWRSGKQREDIKLKDIEHALSVSVFNNKNSGFWHSSLIDRNLIDYFVPFLPLEYKHLKMCIRVEMQSRGYEIDEDIVSRV | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 1861 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction