missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Torsin 1A Monoclonal Antibody (583CT1.1.3)
GREENER_CHOICE

Product Code. 17127178 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17127178 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17127178 Supplier Invitrogen™ Supplier No. MA537587

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Torsin 1A Monoclonal Antibody for Western Blot

The protein encoded by this gene is a member of the AAA family of adenosine triphosphatases (ATPases), is related to the Clp protease/heat shock family and is expressed prominently in the substantia nigra pars compacta. Mutations in this gene result in the autosomal dominant disorder, torsion dystonia 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Torsin 1A
Applications Western Blot
Classification Monoclonal
Clone 583CT1.1.3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.09% sodium azide; pH 7.4
Gene Tor1a
Gene Accession No. O14656
Gene Alias DQ2; dystonia 1; Dystonia 1 protein; Dystonia 1 torsion; dystonia 1, torsion (autosomal dominant; dystonia 1, torsion (autosomal dominant; torsin A); DYT1; LOW QUALITY PROTEIN: torsin-1A; TA; Tor1a; TORA; Torsin 1A; torsin A; torsin ATPase 1; Torsin ATPase-1A; torsin family 1 member A; torsin family 1, member A; torsin family 1, member A (torsin A); torsin-1A; torsinA
Gene Symbols Tor1a
Host Species Mouse
Immunogen purified His-tagged DYT1 protein (fragment).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1861
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.