missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Tornado™ Gas Cell 20m MZFV
Click to view available options
Type:
Tornado™ Gas Cell 20m MZFV
Description
This is an ambient condition long pathlength gas cell with metal body, ZnSe windows, fixed pathlength and Viton O-rings. The pathlength ranges from 2m - 20m in 2m steps and can be used on all FTIR instruments.
Specifications
Specifications
| Type | Tornado™ Gas Cell 20m MZFV |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction