missing translation for 'onlineSavingsMsg'
Learn More

Torkinib, MedChemExpress

Product Code. 16474996
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474996 100 mg 100mg
16470347 50 mg 50mg
16446707 5 mg 5mg
16423998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474996 Supplier Medchem Express Supplier No. HY10474
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Torkinib (PP 242) is a selective and ATP-competitive mTOR inhibitor with an IC50 of 8 nM. PP242 inhibits both mTORC1 and mTORC2 with IC50s of 30 nM and 58 nM, respectively.

Torkinib (PP 242) is a selective and ATP-competitive mTOR inhibitor with an IC50 of 8 nM. PP242 inhibits both mTORC1 and mTORC2 with IC50s of 30 nM and 58 nM, respectively.

Specifications

Chemical Name or Material Torkinib
CAS 1092351-67-1
Color Brown
Purity Grade Notes Research
Molecular Formula C16H16N6O
Quantity 100 mg
Synonym PP 242
Solubility Information DMSO : 50 mg/mL (162.16 mM; Need ultrasonic)
SMILES NC1=C2C(N(N=C2C3=CC4=C(N3)C=CC(O)=C4)C(C)C)=NC=N1
Molecular Weight (g/mol) 308.34
Formula Weight 308.34
Percent Purity 97.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.