missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Torin 2

Description
Potent and selective mTOR inhibitor (IC50 = 2.1 nM). Displays 800-fold cellular selectivity for mTOR over PI3K (cellular EC50 values are 0.25 and 200 nM for mTOR and PI3K respectively). Activates autophagy. Orally available.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 432.4 |
| Purity | >98% |
| Molecular Weight (g/mol) | 432.406 |
| Chemical Name or Material | Torin 2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?