missing translation for 'onlineSavingsMsg'
Learn More

Torin 2, MedChemExpress

Product Code. 16438187
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438187

Brand: Medchem Express HY13002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Torin 2 is an mTOR inhibitor with EC50 of 0.25 nM for inhibiting cellular mTOR activity, and exhibits 800-fold selectivity over PI3K (EC50: 200 nM). Torin 2 also inhibits DNA-PK with an IC50 of 0.5 nM in the cell free assay. Torin 2 can suppress both mTORC1 and mTORC2.

Torin 2 is an mTOR inhibitor with EC50 of 0.25 nM for inhibiting cellular mTOR activity, and exhibits 800-fold selectivity over PI3K (EC50: 200 nM). Torin 2 also inhibits DNA-PK with an IC50 of 0.5 nM in the cell free assay. Torin 2 can suppress both mTORC1 and mTORC2.

Specifications

Chemical Name or Material Torin 2
CAS 1223001-51-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H15F3N4O
Quantity 5 mg
Solubility Information DMSO : 15.62 mg/mL (36.12 mM; Need ultrasonic)
SMILES O=C1N(C2=C(C=C1)C=NC3=CC=C(C=C32)C4=CC=C(N=C4)N)C5=CC=CC(C(F)(F)F)=C5
Molecular Weight (g/mol) 432.4
Formula Weight 432.4
Percent Purity 98.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.