missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Torin 2, MedChemExpress
Description
Torin 2 is an mTOR inhibitor with EC50 of 0.25 nM for inhibiting cellular mTOR activity, and exhibits 800-fold selectivity over PI3K (EC50: 200 nM). Torin 2 also inhibits DNA-PK with an IC50 of 0.5 nM in the cell free assay. Torin 2 can suppress both mTORC1 and mTORC2.
Specifications
Specifications
| Chemical Name or Material | Torin 2 |
| CAS | 1223001-51-1 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H15F3N4O |
| Quantity | 100 mg |
| Solubility Information | DMSO : 15.62 mg/mL (36.12 mM; Need ultrasonic) |
| SMILES | O=C1N(C2=C(C=C1)C=NC3=CC=C(C=C32)C4=CC=C(N=C4)N)C5=CC=CC(C(F)(F)F)=C5 |
| Molecular Weight (g/mol) | 432.4 |
| Formula Weight | 432.4 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?