missing translation for 'onlineSavingsMsg'
Learn More

Torin 2, MedChemExpress

Product Code. 16496056
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496056 100 mg 100mg
16485468 10 mg 10mg
16471056 200 mg 200mg
16461647 50 mg 50mg
16438187 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496056 Supplier Medchem Express Supplier No. HY13002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Torin 2 is an mTOR inhibitor with EC50 of 0.25 nM for inhibiting cellular mTOR activity, and exhibits 800-fold selectivity over PI3K (EC50: 200 nM). Torin 2 also inhibits DNA-PK with an IC50 of 0.5 nM in the cell free assay. Torin 2 can suppress both mTORC1 and mTORC2.

Torin 2 is an mTOR inhibitor with EC50 of 0.25 nM for inhibiting cellular mTOR activity, and exhibits 800-fold selectivity over PI3K (EC50: 200 nM). Torin 2 also inhibits DNA-PK with an IC50 of 0.5 nM in the cell free assay. Torin 2 can suppress both mTORC1 and mTORC2.

Specifications

Chemical Name or Material Torin 2
CAS 1223001-51-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H15F3N4O
Quantity 100 mg
Solubility Information DMSO : 15.62 mg/mL (36.12 mM; Need ultrasonic)
SMILES O=C1N(C2=C(C=C1)C=NC3=CC=C(C=C32)C4=CC=C(N=C4)N)C5=CC=CC(C(F)(F)F)=C5
Molecular Weight (g/mol) 432.4
Formula Weight 432.4
Percent Purity 98.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.