missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Torin 2

Product Code. 17149981
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. Quantity unitSize
17149981 10 mg 10mg
17139981 5 mg 5mg
2 options
This item is not returnable. View return policy

Product Code. 17149981

Brand: STEMCELL Technologies 1000260

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

mTOR pathway inhibitor; Inhibits mTOR

  • Torin 2 is a potent and selective ATP-competitive inhibitor against mTOR (EC50 = 0.25 nM), a key regulator of cell growth, survival, and autophagy (Liu et al.; Petherick et al.)
  • Inhibits proliferation of lung, breast, colorectal, and cervical cancer cell lines (Liu et al.)
  • Blocks mTOR complex 1 (mTORC1)-associated cell cycle progression and induces autophagy in hepatocellular carcinoma cells (Wang et al.)

Specifications

CAS 1223001-51-1
Quantity 10 mg
Molecular Formula C24H15F3N4O
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.