missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Torin 2

Product Code. 17139981
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17139981 5 mg 5mg
17149981 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17139981 Supplier STEMCELL Technologies Supplier No. 1000259

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

mTOR pathway inhibitor; Inhibits mTOR

  • Torin 2 is a potent and selective ATP-competitive inhibitor against mTOR (EC50 = 0.25 nM), a key regulator of cell growth, survival, and autophagy (Liu et al.; Petherick et al.)
  • Inhibits proliferation of lung, breast, colorectal, and cervical cancer cell lines (Liu et al.)
  • Blocks mTOR complex 1 (mTORC1)-associated cell cycle progression and induces autophagy in hepatocellular carcinoma cells (Wang et al.)

Specifications

CAS 1223001-51-1
Quantity 5 mg
Molecular Formula C24H15F3N4O
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.