missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Torin 1
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
- Torin 1 is an ATP-competitive inhibitor of the mammalian target of rapamycin (mTOR), inhibiting the mTOR complexes mTORC1 and mTORC2 with IC50 values of 2 and 10 nM respectively
- It is selective for mTOR versus a panel of over 400 other kinases screened at 10 μM (Thoreen et al.; Liu et al.)
- Inhibits tumor growth, likely through a cytostatic rather than cytotoxic mechanism, in a U87MG PTEN-null glioblastoma mouse xenograft model (Liu et al.)
- Hinders growth, motility, invasion, and survival of primary colon cancer stem-like cells in vitro, and suppresses tumor growth but does not affect normal colon stem cells in xenografts (Francipane & Lagasse)
Specifications
Specifications
| CAS | 1222998-36-8 |
| Quantity | 50 mg |
| Molecular Formula | C35H28F3N5O2 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction