missing translation for 'onlineSavingsMsg'
Learn More

Torcetrapib, MedChemExpress

Product Code. 16434988
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434988

Brand: Medchem Express HY12089

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Torcetrapib (CP-529414) is a selective, potent cholesteryl ester transfer protein (CETP) inhibitor. A typical inhibition curve for whole human plasma, having a CETP concentration of 37 nM.

Torcetrapib (CP-529414) is a selective, potent cholesteryl ester transfer protein (CETP) inhibitor. A typical inhibition curve for whole human plasma, having a CETP concentration of 37 nM.

Specifications

Chemical Name or Material Torcetrapib
Physical Form Solid
CAS 262352-17-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C26H25F9N2O4
Synonym CP-529414
Solubility Information DMSO : ≥ 100 mg/mL (166.54 mM)
SMILES O=C(N1[C@H](CC)C[C@H](N(CC2=CC(C(F)(F)F)=CC(C(F)(F)F)=C2)C(OC)=O)C3=C1C=CC(C(F)(F)F)=C3)OCC
Molecular Weight (g/mol) 600.47
Formula Weight 600.47
Percent Purity 99.39%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.