missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TORC3/CRTC3 Antibody (JE63-18), Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-32226
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TORC3/CRTC3 Monoclonal antibody specifically detects TORC3/CRTC3 in Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
| TORC3/CRTC3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 | |
| CREB regulated transcription coactivator 3, FLJ21868, TORC-3, TORC3CREB-regulated transcription coactivator 3, Transducer of CREB protein 3, transducer of regulated cAMP response element-binding protein (CREB) 3, Transducer of regulated cAMP response element-binding protein 3, transducer of regulated CREB protein 3 | |
| Synthetic peptide within Human TORC3/CRTC3 aa 570-219/ 619. (Uniprot: Q6UUV7) | |
| 100 μL | |
| Primary | |
| Human, Monkey, Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE63-18 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 64784 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido