missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Torc2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534547
This item is not returnable.
View return policy
Description
A suggested positive control is human small intestine tissue lysate. PA5-34547 can be used with blocking peptide PEP-1590.
CRTC2, also known as TORC2 (Transducers Of Regulated cAMP Response Element-Binding (CREB)) 2 and the related protein CRTC1 are potent CREB coactivators that are exported from the nucleus in a CRM1-dependent manner via phosphorylation-dependent interactions. Studies suggest that their phosphorylation and nuclear/cytoplasmic shuttling play an important role in the regulation of gluconeogenesis by cAMP. CRTC2 is present in both B and T-lymphocytes and abundantly expressed in the thymus. Its activity is important in regulating the expression of genes involved in cellular energy metabolism while CRTC1 is essential for energy balance and fertility.
Specifications
| Torc2 | |
| Polyclonal | |
| Unconjugated | |
| CRTC2 | |
| 4632407F12Rik; CREB regulated transcription coactivator 2; CREB-regulated transcription coactivator 2; CRTC2; mTORC2; OTTHUMP00000035255; RGD1308903; TORC2; TORC-2; transducer of CREB protein 2; transducer of regulated cAMP response element-binding protein (CREB) 2; transducer of regulated cAMP response element-binding protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200186, 310615, 74343 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q3LRZ1, Q3U182, Q53ET0 | |
| CRTC2 | |
| A 16 amino acid peptide near the carboxy terminus of human CRTC2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction